missing translation for 'onlineSavingsMsg'
Learn More

HPS1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

136.48 CHF - 385.92 CHF

Spezifikation

Antigen HPS1
Konzentration 0.24 mg/mL
Anwendungen Immunofluorescence, Immunocytochemistry, Western Blot
Klassifikation Polyclonal
Konjugat Unconjugated
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Product Code Brand Menge Price Quantity & Availability  
16864074
missing translation for 'viewDocuments'
Proteintech
15077-1-AP-20UL
20 μL
136.48 CHF
20 Mikroliter
View alternative products
missing translation for 'toSaveUpto' 
»
om dit product te kopen Registreer vandaag om een webaccount aan te maken
16854074
missing translation for 'viewDocuments'
Proteintech
15077-1-AP-150UL
150 μL
385.92 CHF
150 Mikroliter
View alternative products
missing translation for 'toSaveUpto' 
»
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Description

Description

Hermansky-Pudlak syndrome (HPS) is a rare, genetically heterogeneous, autosomal recessive disorder. It is characterized by oculocutaneous albinism, lysosomal storage defects and prolonged bleeding due to platelet storage pool deficiency. There are 10 HPS genes encoding HPS proteins that all interact within three distinct ubiquitously expressed protein complexes or biogenesis of lysosome-related organelle complexes. Defects in these genes cause HPS. HSP-1 is a component of multiple cytoplasmic organelles and is important for their normal development and function. It plays a role in intracellular protein sorting.
Specifications

Specifications

HPS1
Immunofluorescence, Immunocytochemistry, Western Blot
Unconjugated
Rabbit
Human
Q92902
3257
HPS1 Fusion Protein Ag7149
Primary
-20°C
HPS1
0.24 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Hermansky Pudlak syndrome 1, HPS, HPS1
HPS1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.