missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPDL Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20777-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
May have dioxygenase activity.Spezifikation
HPDL | |
Polyclonal | |
Unconjugated | |
HPDL | |
4 HPPD L, GLOXD1, HPDL, RP4 534D1.1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
84842 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96IR7 | |
HPDL | |
HPDL Fusion Protein Ag14773 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |