missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hnRNP M1-M4 Antibody (1D8), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 6 publications
Marke: Novus Biologicals NB200-314
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
hnRNP M1-M4 Monoclonal specifically detects hnRNP M1-M4 in Human, Mouse, Rat, Porcine, Bovine, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Spezifikation
| hnRNP M1-M4 | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.1% Sodium Azide | |
| DKFZp547H118, heterogeneous nuclear ribonucleoprotein M, hnRNP M, HNRNPM4, HNRPM4heterogenous nuclear ribonucleoprotein M, HNRPMheterogenous nuclear ribonucleoprotein M4, HTGR1, M4 protein, N-acetylglucosamine receptor 1, NAGR1hnRNA-binding protein M4 | |
| Mouse | |
| Unpurified | |
| RUO | |
| Primary | |
| NB200-314 is specific for all four M proteins of hnRNP (M1, M2, M3 and M4). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 1D8 | |
| Western Blot 1:500, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen 1:10-1:500 | |
| P52272 | |
| HNRNPM | |
| M19 fusion protein containing full-length human protein. [UniProt# P52272] | |
| 0.2 mL | |
| Cancer | |
| 4670 | |
| Human, Mouse, Rat, Porcine, Bovine, Rabbit | |
| Ascites |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur