missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGN4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11686-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Description
HMGN4, also named as High mobility group nucleosome-binding domain-containing protein 4, is a 90 amino acid protein, which belongs to the HMGN family. HMGN4 as a DNA-binding protein localizes in the nucleus. The calculated molecular weight of HMGN4 is 10 kDa, but the modified HMGN4 protein is about 15 kDa.Specifications
HMGN4 | |
Polyclonal | |
Unconjugated | |
HMGN4 | |
HMG17L3, HMGN4, NHC | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10473 | |
Antibody | |
IgG |
Immunofluorescence, Immunocytochemistry, Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O00479 | |
HMGN4 | |
HMGN4 Fusion Protein Ag2298 | |
20 μL | |
Primary | |
Human | |
Liquid |