missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIV1 p24 Polyclonal Antibody

Goat Polyclonal Antibody
Marke: Invitrogen™ PA17217
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
PA1-7217 detects the HIV1 p24. This antibody does not cross-react with human T or B cells. PA1-7217 has been successfully used in Western blot procedures. The PA1-7217 immunogen is purified native p24 from strain IIIB.
HIV-1 Gag p24 is a capsid protein that constitutes the core of the AIDS virus HIV-1 and is produced by the digestion of its precursor Gag p55 by HIV-1 protease. The HIV-1 Gag p24 protein is indispensable to the reproduction of AIDS virus, and constitutes an essential element for the AIDS virus particle construction. HIV-1 Gag p24 is detectable from the early stage of AIDS virus infection, and is used as a marker for observation of the development in the patient's condition after treatment, as it indicated the amount of virus in the blood.
Spezifikation
| HIV1 p24 | |
| Polyclonal | |
| Unconjugated | |
| HIV p24; HIV1gp1 | |
| Purified native p24 from strain IIIB. | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 5 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Goat | |
| IgG fraction | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur