missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIV1 p24 Monoclonal Antibody (D45F), FITC

Mouse Monoclonal Antibody
Marke: Invitrogen™ MA17378
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The MA1-7378 antibody reacts with HIV-1 p24 in viral and infected tissue samples. MA1-7378 has been successfully used in ELISA, Immunofluorescence, Western blot and Immunohistochemical applications. The MA1-7378 antibody was raised against HIV protein 24.
HIV-1 Gag p24 is a capsid protein that constitutes the core of the AIDS virus HIV-1 and is produced by the digestion of its precursor Gag p55 by HIV-1 protease. The HIV-1 Gag p24 protein is indispensable to the reproduction of AIDS virus, and constitutes an essential element for the AIDS virus particle construction. HIV-1 Gag p24 is detectable from the early stage of AIDS virus infection, and is used as a marker for observation of the development in the patient's condition after treatment, as it indicated the amount of virus in the blood.
Spezifikation
| HIV1 p24 | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry, Western Blot | |
| D45F | |
| FITC | |
| HIV p24; HIV1gp1 | |
| HIV protein 24. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur