missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIV1 p24 Monoclonal Antibody (5)
Mouse Monoclonal Antibody
Marke: Invitrogen™ MA171515
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This antibody reacts with HIV-1 p24 subtype B, C and A, and has been mapped to amino acids 48-62 (TPQDLNTMLNTVGGH). This clone can also be referred to as 5 (05-001).
HIV-1 Gag p24 is a capsid protein that constitutes the core of the AIDS virus HIV-1 and is produced by the digestion of its precursor Gag p55 by HIV-1 protease. The HIV-1 Gag p24 protein is indispensable to the reproduction of AIDS virus, and constitutes an essential element for the AIDS virus particle construction. HIV-1 Gag p24 is detectable from the early stage of AIDS virus infection, and is used as a marker for observation of the development in the patient's condition after treatment, as it indicated the amount of virus in the blood.
Spezifikation
| HIV1 p24 | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.4 | |
| Mouse | |
| Protein G | |
| RUO | |
| Virus | |
| Antibody | |
| IgG2a |
| ELISA, Western Blot | |
| 5 | |
| Unconjugated | |
| HIV p24; HIV1gp1 | |
| Recombinant p24 protein of HIV-1 HAN consensus purified from E. coli. | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur