Learn More
Invitrogen™ Histone H3 Peptide (1-20)
Matched and validated lipid and peptide substrates are available to complement the Adapta™ Universal Kinase Assay.
Marke: Invitrogen™ PV5707
Additional Details : Gewicht : 0.00100kg
Beschreibung
Adapta™ Universal Kinase assays developed with these reagents allow for primary or secondary screening of kinase inhibitors.
Spezifikation
Kinase Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Pharma and Biopharma, Target and Lead Identification and Validation | |
Substrates for Adapta™ Assays are shipped on dry ice and should be stored at -20°C. | |
ARTKQTARKSTGGKAPRKQLW |
1mL | |
RUO - research use only | |
Liquid |