missing translation for 'onlineSavingsMsg'
Learn More

HIRIP3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Artikelnummer. 16823954 Alle ansehen Proteintech Produkte
Change view
Click to view available options
Menge:
20 μL
150 μL
Packungsgröße:
150 Mikroliter
20 Mikroliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
16823954 20 μL 20 Mikroliter
16813954 150 μL 150 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 16823954 Lieferant Proteintech Lieferanten-Nr. 149921AP20UL

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

The HIRA protein shares sequence similarity with Hir1p and Hir2p, the two corepressors of histone gene transcription characterized in the yeast, Saccharomyces cerevisiae. The structural features of the HIRA protein suggest that it may function as part of a multiprotein complex. Several cDNAs encoding HIRA-interacting proteins, or HIRIPs, have been identified. In vitro, the protein encoded by this gene binds HIRA, as well as H2B and H3 core histones, indicating that a complex containing HIRA-HIRIP3 could function in some aspects of chromatin and histone metabolism. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.

Specifications

Antigen HIRIP3
Anwendungen Western Blot, Immunohistochemistry (Paraffin)
Klassifikation Polyclonal
Konzentration 0.13 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gen HIRIP3
Gen-Zugriffsnummer Q8BLH7, Q9BW71
Gen-Alias HIRA interacting protein 3, HIRIP3
Gensymbole HIRIP3
Wirtsspezies Rabbit
Immunogen HIRIP3 Fusion Protein Ag6978
Reinigungsverfahren Antigen Affinity Chromatography
Menge 20 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 233876, 8479
Zielspezies Human, Mouse
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.