Learn More
Tocris Bioscience™ Hh-Ag1.5
Click to view available options
Menge:
1 mg
5 mg
Packungsgröße:
1 Milligramm
5 Milligramm
Beschreibung
Hh-Ag1.5 is a potent and high affinity Hedgehog (Hh) receptor agonist (EC50 = 1 nM; Ki values are 0.5 and 2.3 nM respectively, in a [3H]SAG-1.3 and [3H]Cyclopamine binding assay). It is used in protocols for the chemical reprogramming of mouse embryonic fibroblasts into neural stem cells. Also induces differentiation of hiPSCs into skin-derived precursor cells, spinal motor neurons and spinal sensory neurons.
Spezifikation
Spezifikation
| Chemischer Name oder Material | 3-Chloro-4,7-difluoro-N-[trans-4-(methylamino)cyclohexyl]-N-[[3-(4-pyridinyl)phenyl]methyl]benzo[b]thiophene-2-carboxamide |
| CAS | 612542-14-0 |
| Menge | 5 mg |
| Ziel | Smoothened |
| Summenformel | C28H26ClF2N3OS |