missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Herpes Simplex Virus VP16 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals (Bio-Techne) NB600-950CL1
This item is not returnable.
View return policy
Description
Herpes Simplex Virus VP16 Polyclonal antibody specifically detects Herpes Simplex Virus VP16 in Virus samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| Herpes Simplex Virus VP16 | |
| Polyclonal | |
| PBS | |
| Synthetic peptide corresponding to amino acids 474-487 of the herpes simplex virus Herpes Simplex Virus VP16 protein, conjugated to KLH via an N-terminal added cysteine. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction