missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Heratherm Incubation Chambers

Artikelnummer. 31926567 Alle ansehen Thermo Scientific Produkte
Ansicht ändern
Click to view available options
Ausführung der Tür:
Glas
Glass door
Massiv (außen), Glas (innen)
Solid outer door with inner glass door
Kapazität (metrisch):
247 L
750 L
Packungsgröße:
1 Einheit
1 Stück
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Ausführung der Tür Kapazität (metrisch) unitSize
31926567 Glass door 247 L 1 Stück
18794584 Massiv (außen), Glas (innen) 750 L 1 Einheit
18774584 Glas 750 L 1 Einheit
31926573 Solid outer door with inner glass door 247 L 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 31926567 Lieferant Thermo Scientific™ Lieferanten-Nr. EIG250
 Eingeschränkter Artikel
Verfügbar ab: 03-09-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Comprehensive data monitoring and temperature control combined with a flexible Heratherm Incubation Chamber provide a dependable option for microbiological incubation or temperature-dependent storage and testing.

Produces precise temperature conditions above and below ambient, ideally suited for microbiological incubation, and heated or cooled incubated storage.

  • Uniform Temperature throughout the chamber.
  • Energy-efficient Peltier technology supports temperature stability and uniformity with lower energy usage compared to traditional compressors and without hazardous refrigerant.
  • Intuitive touchscreen interface with easy programmability of temperature cycles and process duration.
  • Two access ports for independent data capture, and internal documentation of set and actual parameter values, along with event log to meet 21 CFR Part 11 requirements for process documentation.
  • Data retrieval via USB stick, or Thermo Cloud via OPC-UA standard.
  • Sample and process protection: Assign personalized access with user IDs; restrict access with standard door lock, or optional automated door lock; various alarm options to prevent operator of process issues.
  • Simplified setup with no technician required; install using standard power cord.
  • IQ/OQ qualifications and other services available; visit thermofisher.com/chamberservice.
  • Inner glass door keeps environment stable, even when opening outer door. Glass door also available
  • Rounded corners and removable shelving offer easy cleaning to help prevent cross-contamination.
  • Interior electrical socket for use of stirrer, shaker or other equipment inside the chamber are available as a factory-installed option. Choose from a variety of country-specific sockets. (Ordered with separate order code)
TRUSTED_SUSTAINABILITY

Spezifikation

Alarmtypen Audible and Visible
Kapazität (angloamerikanisch) 8.8 cu.ft.
Fassungsvermögen (angloamerikanisch) Einschub 66.1 lb.
Fassungsvermögen (metrisch) Einschub 30 kg
Zertifizierung/Konformität UL, CE
Datenausgabe USB, Thermo Cloud via OPC UA
Kapazität (metrisch) 247 L
Kammermaterial Powder-coated Cold Rolled Steel
Beleuchtung No
Material Powder-coated Cold Rolled Steel
Anzahl Einschübe 2
Steckertyp Nema 5-20P
Relative Luftfeuchtigkeit 10 to 90%
Temperaturbereich (metrisch) 0 to 70°C
Typ Temperature Test Chamber
Display 7 in. Touchscreen user interface
Ausführung der Tür Glass door
Zur Verwendung mit (Anwendung) Incubation, storage, temperature testing
Anzahl Rollen None
Anschlüsse 2 access ports
Programmierbar Temperature ramping
Einschubfläche (angloamerikanisch) 3.05 ft.2
Einschubfläche (metrisch) 0.28 m2
Einschubmaterial Stainless Steel (304/1.4301)
Temperaturregelung Microprocessor - programmable
Temperaturauflösung 0.1°C
Temperaturstabilität ≤ ±0.1°C @ 20°C to 37°C
Temperaturgleichförmigkeit ≤ ±0.5°C @ 20°C to 37°C
Gewährleistung 24 months
Breite (angloamerikanisch) 35.5 in.
Innenbreite (angloamerikanisch) 26 in.
Breite (angloamerikanisch) Einschub 24.2 in.
Breite (metrisch) 901 mm
Innenbreite (metrisch) 660 mm
Breite (metrisch) Einschub 615 mm
Höhe (angloamerikanisch) 44.4 in.
Innenhöhe (angloamerikanisch) 31.5 in.
Höhe (metrisch) 1128 mm
Innenhöhe (metrisch) 800 mm
Tiefe (angloamerikanisch) 31.1 in.
Innentiefe (angloamerikanisch) 20.4 in.
Tiefe (metrisch) 790 mm
Innentiefe (metrisch) 519 mm
Versandgewicht (angloamerikanisch) 448 lb.
Versandgewicht (metrisch) 203 kg
Elektrische Anforderungen 100/240V, 50/60 Hz
Spannung 100/115/220/230 V
Wirkleistung 600 W
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.