missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Hemagglutinin-Neuraminidase Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA5109232
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Positive Control: Hemagglutinin-neuraminidase transfected HEK293 cell.
Hemagglutinin-neuraminidase refers to a single viral protein that has both hemagglutinin and neuraminidase activity. This is in contrast to the proteins found in influenza, where both functions exist but in two separate proteins. However it does show a structural similarity to influenza viral neuraminidase and has a six-bladed beta-propeller structure. Hemagglutinin-neuraminidase allows the virus to stick to a potential host cell, and cut itself loose if necessary. Hemagglutinin-neuraminidase can be found in a variety of paramyxoviruses including mumps virus, human parainfluenza virus 3, and the avian pathogen Newcastle disease virus.
Spezifikation
| Hemagglutinin-Neuraminidase | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| Synthetic peptide within newcastle disease virus Hemagglutinin-neuraminidase aa 537-586. | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur