Learn More
Heidolph™ Hei-VAP Core All-round Chill Package, CH-plug
Artikelnummer. 17353490 Alle ansehen Heidolph Instruments Produkte
Ansicht ändern
Click to view available options
Steckertyp:
CH-Stecker
Packungsgröße:
1 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Steckertyp Packungsgröße
17353490 CH-Stecker 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17353490 Lieferant Heidolph™ Lieferanten-Nr. 5712130004
Verfügbar ab: 02-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
The Hei-VAP University Academic Package features a hand lift, vacuum pump, chiller, and tubing set.
  • Hei-VAP Core with hand lift and vertical glassware set G3
  • Hei-VAC Control
  • Vacuum pump Rotavac Valve Tec
  • Tubing set
  • Chiller Hei-CHILL 250
  • Thermal fluid Kyro 30 (10 L)
TRUSTED_SUSTAINABILITY
Spezifikation
Typ Hand Lift Rotary Evaporator
Steckertyp CH-Stecker