Learn More
Heidolph™ Hei-VAP Advanced Evaporation Package, CH-plug
Artikelnummer. 17303510 Alle ansehen Heidolph Instruments Produkte
Ansicht ändern
Click to view available options
Steckertyp:
CH-Stecker
Packungsgröße:
1 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Steckertyp unitSize
17303510 CH-Stecker 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17303510 Lieferant Heidolph™ Lieferanten-Nr. 5723630004
Verfügbar ab: 02-09-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
The Hei-VAP Advanced Evaporation Package features a motor lift, vacuum pump, and tubing set.
  • Hei-VAP Core with motor lift and vertical glassware set G3
  • Vacuum pump Rotavac Vario Pumping Unit
  • Tubing set
TRUSTED_SUSTAINABILITY
Spezifikation
Typ Motor Lift Rotary Evaporator
Steckertyp CH-Stecker