Learn More
Heidolph™ Hei-FLOW Ultimate 120 Multi
Artikelnummer. 17353450 Alle ansehen Heidolph Instruments Produkte
Ansicht ändern
Click to view available options
Steckertyp:
CH-Stecker
Packungsgröße:
1 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Steckertyp Packungsgröße
17353450 CH-Stecker 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17353450 Lieferant Heidolph™ Lieferanten-Nr. 5235201303
Verfügbar ab: 29-10-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Model that allows you to choose between a single-channel or multi-channel pump head.
  • Accurate dosing is particularly important in the pharmaceutical, cosmetic or food sector. Comprehensible process conditions are a prerequisite for the safe conveyance of sensitive media. This is ensured by a control accuracy of ±1% and precise speed adjustment (5 to 120 rpm).
  • In addition, an optional foot switch allows the pump to be operated in closed exhaust hoods. The parameters - speed, direction of rotation and on/off function - can be controlled externally either through the analog or digital interface.
  • In this model, you can choose between a single-channel or multichannel pump head. Flow rates vary depending on the pump head: the single-channel head has a flow rate of 0.38 to 813 mL/min; for the multichannel head 0.005 to 364 mL/min. Depending on the focus of the process, the following parameters can be set on the device: Speed, tubing diameter, flow rate display, metering volume, interval dosing and break times. The pump can be calibrated for optimum dosing results.
TRUSTED_SUSTAINABILITY
Spezifikation
Regelungsart Digitale variable Drehzahl
Displaytyp Analog
Schutzart Gehäuse IP55
Wandstil Variable-Speed Digital
Gewicht (angloamerikanisch) IP 55
Spannung 230 V
Wirkleistung Analog
Steckertyp CH-Stecker