Learn More
Axygen™ Heating Block
Alle ansehen Corning Produkte
Click to view available options
Röhrchenkapazität:
0,2 ml
0,5 mL
1,5 ml
2 mL
5 ml
15 ml
50 ml
Wellanzahl:
24
4
8
96
96/384
Modell:
Flachboden
Konusboden
Rundboden
Packungsgröße:
1 Stück
Beschreibung
- No additional tools are required for block exchanges
- The blocks utilize a magnetic adhesion technology and are self-recognized once exchanged
- The combination of heating, cooling, and mixing with a versatile block selection makes the Axygen Microtube Shaker ideal for many life science laboratories in the segments of molecular biology, biochemistry, and clinical chemistry
- Applications include but are not limited to gene synthesis, gene purification and denaturation, enzymatic reactions, and bacterial growth assays
Spezifikation
Spezifikation
| Röhrchenkapazität | 50 ml |
| Gewicht (metrisch) | 0.3 kg |
| Modell | Flachboden |
| Wellanzahl | 4 |