Learn More
Axygen™ Heating Block
Artikelnummer. 17802617 Alle ansehen Corning Produkte
Click to view available options
Röhrchenkapazität:
0,2 ml
0,5 mL
1,5 ml
2 mL
5 ml
15 ml
50 ml
Wellanzahl:
24
4
8
96
96/384
Modell:
Flachboden
Konusboden
Rundboden
Packungsgröße:
1 Stück
Artikelnummer. 17802617
Marke: Axygen™ I4010I
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Interchangeable aluminum block to accommodate PCR plates and a variety of common life science vessels.
  • No additional tools are required for block exchanges
  • The blocks utilize a magnetic adhesion technology and are self-recognized once exchanged
  • The combination of heating, cooling, and mixing with a versatile block selection makes the Axygen Microtube Shaker ideal for many life science laboratories in the segments of molecular biology, biochemistry, and clinical chemistry
  • Applications include but are not limited to gene synthesis, gene purification and denaturation, enzymatic reactions, and bacterial growth assays
TRUSTED_SUSTAINABILITY
Spezifikation
Röhrchenkapazität 0,2 ml
Gewicht (metrisch) 0.3 kg
Modell Flachboden
Wellanzahl 96/384