Learn More
ACROBiosystems HCMV Glycoprotein B (gB) Protein, Fc Tag
Marke: ACROBiosystems CMB-V5255-500UG
Additional Details : Gewicht : 0.00001kg
Beschreibung
Human cytomegalovirus is a species of the Cytomegalovirus genus of viruses, which in turn is a member of the viral family known as Herpesviridae or herpesviruses. It is typically abbreviated as HCMV or, commonly but more ambiguously, as CMV. CMV Virus Envelope Glycoportein B (CMV-GB) can be cleaved into glycoprotein GP55. Envelope glycoprotein that plays a role in host cell entry, cell to-cell virus transmission, and fusion of infected cells. CMV-GB may be involved in the initial attachment via binding to heparan sulfate together with the gM/gN complex that binds heparin with higher affinity. Furthermore, CMV-GB can interact with host integrin ITGB1, PDGFRA and EGFR that likely serve as postattachment entry receptors. Also, CMV-GB participates in the fusion of viral and cellular membranes leading to virus entry into the host cell. Membrane fusion is mediated by the fusion machinery composed at least of gB and the heterodimer gH/gL.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 104.0 kDa. The protein migrates as 130-180 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAA45920.1 | |
500 μg | |
Val 23 - Lys 700 | |
Glycoprotein B,gB,Envelope glycoprotein B | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
104.0 kDa | |
HCMV Glycoprotein B (gB), Fc Tag (CMB-V5255) Val 23 - Lys 700 (Accession # P13201-1) with furin cleavage site mutated from 'RTKR' to 'TTQT', was produced in human 293 cells (HEK293) (at ACROBiosystems.) | |
-20°C | |
Glycoprotein B / gB | |
HCMV | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Viral Proteins |