missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HBQ1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 19997-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The antibody is specific to HBQ1.
Theta-globin mRNA is found in human fetal erythroid tissue but not in adult erythroid or other nonerythroid tissue. The theta-1 gene may be expressed very early in embryonic life, perhaps sometime before 5 weeks. Theta-1 is a member of the human alpha-globin gene cluster that involves five functional genes and two pseudogenes. The order of genes is: 5′ - zeta - pseudozeta - mu - pseudoalpha-2 -pseudoalpha-1 - alpha-2 - alpha-1 - theta-1 - 3′. Research supports a transcriptionally active role for the gene and a functional role for the peptide in specific cells, possibly those of early erythroid tissue.Spezifikation
HBQ1 | |
Polyclonal | |
Unconjugated | |
HBQ1 | |
HBQ1, Hemoglobin subunit theta 1, Hemoglobin theta 1 chain, hemoglobin, theta 1, Theta 1 globin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3049 | |
-20°C | |
Liquid |
Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P09105 | |
HBQ1 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |