missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HB9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA540456
Additional Details : Gewicht : 0.00010kg
Beschreibung
Peptide sequence: AAASGTGGGG GGGGASGGTS GSCSPASSEP PAAPADRLRA ESPSPPRLLA Sequence homology: Dog: 86%; Human: 100%; Mouse: 79%; Pig: 93%; Rat: 79%
HB9 Polyclonal antibody specifically detects HB9 in Human samples. It is validated for Western Blot.Spezifikation
HB9 | |
Polyclonal | |
Unconjugated | |
MNX1 | |
HB9; Hlxb9; homeo box HB9; homeobox gene HB9; homeobox HB9; homeobox protein HB9; homeodomain protein Hb9; HOX SCRA1; HOXHB9; LOW QUALITY PROTEIN: motor neuron and pancreas homeobox protein 1; MNR2; MNX1; motor neuron and pancreas homeobox 1; motor neuron and pancreas homeobox protein 1; SCRA1; zgc:112174 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
3110 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P50219 | |
MNX1 | |
Synthetic peptide directed towards the N-terminal of human MNX1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |