missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HA Recombinant Influenza A, subtype H1N1 (A/California/7/2009) Protein, His Tag
HA Recombinant Influenza A, subtype H1N1 (A/California/7/2009) Protein, His Tag
Marke: Invitrogen 11085V08H250
Additional Details : Gewicht : 0.01000kg
Beschreibung
Hemagglutinin (HA), recombinant viral protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of Influenza A virus H1N1 (A/California/07/2009) (ACP44189.1) hemagglutinin (Met 1-Gln 529) (HA1+HA2, two amino acids deletion, uncleaved) fused to a polyhistidine tag at the C-terminus.
Activity: This recombinant protein has not been tested.
Formulation: 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween80.
Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Expiration Date: Expires one year from date of receipt when stored as instructed.
This protein is manufactured by SINO Biological.
Spezifikation
Protein Assays and Analysis, Assay Standard | |
59kDa | |
7.4 ± 0.5 | |
5 x50μg | |
For research use only. Not for use in diagnostic procedures | |
<1 EU/μg | |
HA Recombinant Influenza A Subtype H1N1 (A/California/7/2009) Protein | |
HA Recombinant Influenza A, Subtype H1N1 (A/California/7/2009) Protein | |
His Tag | |
Cell structure/organelle proteins |
Lyophilized | |
HA Recombinant Influenza A Subtype H1N1 (A/California/7/2009) Protein | |
PAGE | |
–20°C | |
1 Each | |
HA | |
HA | |
Human | |
Mammalian | |
Lyophilized |
For Research Use Only. Not for use in diagnostic procedures.