Learn More
Bel-Art™ H-B DURAC Calcium Chloride Salometer
Alle ansehen SP Scienceware Produkte
Click to view available options
Typ:
Salt Hydrometer
Beschreibung
- Choose from two models: one measuring percent saturation of CaCl2 from 0 to 120%, and the other measuring specific gravity/relative density (g/cm³) from 1.000 to 1.280 for versatile testing needs.
- Used across multiple industries including food, fishing, and transportation for reliable brine monitoring.
- 300 mm (12 in.) length for easy handling during solution testing.
- Lead-free ballast and glass construction supports safer laboratory and industrial use.
- Individually serialized for precise identification and quality assurance tracking.
- Temperature standardized at 60°F (15°C) for consistent and accurate readings.
- Hand-inspected by our U.S. quality team to ensure performance and durability.
- Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST for certified measurement confidence.
- Store in a clean, dry, safe place and avoid extreme temperatures to maintain long-term instrument integrity.
Spezifikation
Spezifikation
| Typ | Salt Hydrometer |
| Länge (metrisch) | 30 cm |
| Spannung | 0/1 V |
| Bereich der relativen Dichte | 1.000 to 1.280 |