Learn More
Abnova™ GSS (Human) Recombinant Protein
Click to view available options
Menge:
10 μg
25 μg
Packungsgröße:
10 Mikrogramm
25 Mikrogramm
Beschreibung
- Sequence: MATNWGSLLQDKQQLEELARQAVDRALAEGVLLRTSQEPTSSEVVSYAPFTLFPSLVPSALLEQAYAVQMDFNLLVDAVSQNAAFLEQTLSSTIKQDDFTARLFDIHKQVLKEGIAQTVFLGLNRSDYMFQRSADGSPALKQIEINTISASFGGLASRTPAVHRHVLSVLSKTKEAGKILSNNPSKGLALGIAKAWELYGSPNALVLLIAQEKERNIFDQRAIENELLARNIHVIRRTFEDISEKGSLDQDRRLFVDGQEIAVVYFRDGYMPRQYSLQNWEARLLLERSHAAKCPDIATQLAGTKKVQQELSRPGMLEMLLPGQPEAVARLRATFAGLYSLDVGEEGDQAIAEALAAPSRFVLKPQREGGGNNLYGEEMVQALKQLKDSEERASYILMEKIEPEPFENCLLRPGSPARVVQCISELGIFGVYVRQEKTLVMNKHVGHLLRTKAIEHADGGVAAGVAVLDNPYPV
Spezifikation
Spezifikation
| Zugriffsnummer | AAH07927 |
| Gen-ID (Entrez) | 2937 |
| Name | glutathione synthetase |
| Vorbereitungsmethode | Wheat germ expression system |
| Qualitätskontrollen | 125% SDS-PAGE Stained with Coomassie Blue |
| Menge | 10 μg |
| Lagerungsbedingungen | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Gen-Alias | GSHS, MGC14098 |
| Gensymbol | GSS |
| Spezies | Wheat Germ (in vitro) |
| Mehr anzeigen |