missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSK3B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 22104-1-AP-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This antibody recognize the C-terminal of GSK3B.
Menin is a putative tumor suppressor associated with a syndrome known as multiple endocrine neoplasia type 1. In vitro studies have shown menin is localized to the nucleus, possesses two functional nuclear localization signals, and inhibits transcriptional activation by JunD, however, the function of this protein is not known. Two messages have been detected on northern blots but the larger message has not been characterized. Two variants of the shorter transcript have been identified where alternative splicing affects the CDS. Five variants where alternative splicing takes place in the 5′ UTR have also been identified.Spezifikation
| GSK3B | |
| Polyclonal | |
| Unconjugated | |
| GSK3B | |
| GSK 3 beta, GSK3 beta, GSK3A B, GSK3B, GSK-3B, GSK3B 216P, gsk3beta | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2932, 56637, 84027 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P18266, P49841, Q9WV60 | |
| Gsk3b | |
| GSK3B Fusion Protein Ag17320 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur