missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Griffonia Simplicifolia (African Legume) Lectin I (GSL I), DyLight 594

Artikelnummer. 17106642
Click to view available options
Menge:
0,5 mg
Packungsgröße:
0.50 Milligramm
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 17106642

Marke: Invitrogen™ L32473

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Bright red GSL I-B4-fluorophor conjugate that is sourced from the African legume Griffonia simplicifolia (Bandeiraea simplicifolia)

  • Griffonia Simplicifolia Lectin I (GSL I-B4), DyLight 594, is a bright red GSL I-B4-fluorophor conjugate that is sourced from the African legume Griffonia simplicifolia (Bandeiraea simplicifolia).
  • This B4 isoform of GSL I contains only B subunits and is widely used as a marker of endothelial cells in non-primates such as mouse, rat, rabbit, and goat, as well as a marker of non-peptidergic unmyelinated primary afferent neurons.
  • Very bright DyLight 594 conjugate detected with traditional Texas Red filter set, Ex/Em: 592/617 nm
  • Applications: immunofluorescence, glycobiology
  • Sugar specificity: galactose
  • Inhibiting/eluting sugar: 500 mM galactose (S-9003) or 100 mM raffinose
  • Blood group specificity: B>>A1
  • Unconjugated lectin molecular weight: 114 KDa
  • Immunofluorescence working concentration: 5–20 μg/mL
  • GSL I-B4-DyLight 594 is provided at a 1 mg/mL concentration in 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide, 0.1 mM Ca2+. If a precipitate forms upon long-term storage, warm to 37°C.
  • GSL I is a tetrameric lectin consisting of A and B subunits. There are five GSL-I isolectins with different subunit compositions. The B4 isolectin (all B subunits) is blood-group-B specific and has an exclusive affinity for terminal α-D-galactosyl residues. It specifically agglutinates blood group B erythrocytes and was originally employed for this purpose.
  • Subsequent work has shown that the GSL I B4 isolectin is cytotoxic to several normal and tumor cell types and has particularly strong affinity for brain microglial and perivascular cells. It has also been particularly valuable as a histochemical and flow cytometric probe for specifically labeling endothelial cells in a number of species.
  • GSL I B4 isolectin has been used effectively for tracing central and peripheral neuronal pathways following local injections, as well as for labeling stimulated murine macrophages, bovine thyroid cells, various murine cell types, laminin, and thyroglobulin.
TRUSTED_SUSTAINABILITY

Spezifikation

Menge 0,5 mg
Quelle Bandeiraea Simplicifolia
Konjugat DyLight 594
Inhalt und Lagerung When stored at or below 2–8°C in the dark, the product is stable for 6 months after reciept.

Markertyp DyLight Fluors
Versandbedingung Approved for shipment at Room Temperature or on Wet Ice
Form Solution
Protein-Subtyp Lectins
Produktlinie DyLight

For Research Use Only. Not for use in diagnostic procedures.

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt