Learn More
Granzyme B Mouse anti-Human, Alexa Fluor 647, Clone: GB11, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 560212
Additional Details : Gewicht : 0.01000kg
Beschreibung
The GB11 antibody specifically reacts with human granzyme B, a serine protease of approximately 32 kDa. Granzyme B is stored in the granules of cytotoxic T lymphocytes and NK cells along with the pore-forming protein perforin. In the classic model of target cell lysis, perforins create holes in the target cell membrane allowing entrance of granzymes. Granzyme B has been shown to act on specific substrates including caspase-3, -7, -9, and -10 which in turn give rise to enzymes that mediate apoptosis. Granzyme B may also be involved in the hydrolysis of extracellular matrix components. Detectable levels of granzyme B have been detected in sera from healthy volunteers. The immunogen used to generate the GB11 hybridoma was human granzyme B isolated from an NK cell line.
Spezifikation
Granzyme B | |
Monoclonal | |
0.2mg/mL | |
GZMB; Granzyme-2; CCPI; CGL1; CSPB; CTLA1; CTSGL1; GRB; HLP; SECT | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
GB11 | |
Alexa Fluor 647 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human Granzyme B | |
0.1 mg | |
Immunology | |
Human | |
IgG1 κ |
For Research Use Only.