missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR81 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20146-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The G protein-coupled receptor GPCR GPR81 has been reported primarily in adipose and pituitary. ESTs have been isolated from human bladder cancer and colon cancer libraries. It has not been detected in frontal, temporal and occipital lobes of the cortex, basal forebrain, caudate nucleus, nucleus accumbens, and hippocampus.Spezifikation
GPR81 | |
Polyclonal | |
Unconjugated | |
HCAR1 | |
FKSG80, G protein coupled receptor 104, G protein coupled receptor 81, GPR104, GPR81, LACR1, TA GPCR | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27198 | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin) | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BXC0 | |
HCAR1 | |
GPR81 Fusion Protein Ag13682 | |
20 μL | |
Primary | |
Human | |
Liquid |