missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR171 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20307-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Spezifikation
GPR171 | |
Polyclonal | |
Unconjugated | |
GPR171 | |
G protein coupled receptor 171, GPR171, H963 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29909 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O14626 | |
GPR171 | |
GPR171 Fusion Protein Ag14141 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |