missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPNMB Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20338-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is a type I transmembrane glycoprotein which shows homology to the pMEL17 precursor, a melanocyte-specific protein. GPNMB shows expression in the lowly metastatic human melanoma cell lines and xenografts but does not show expression in the highly metastatic cell lines. GPNMB may be involved in growth delay and reduction of metastatic potential. Two transcript variants encoding different isoforms have been found for this gene.Spezifikation
GPNMB | |
Polyclonal | |
Unconjugated | |
GPNMB | |
GPNMB, HGFIN, Transmembrane glycoprotein NMB | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10457, 93695 | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q14956, Q99P91 | |
Gpnmb | |
GPNMB Fusion Protein Ag14124 | |
20 μL | |
Primary | |
Human, Mouse | |
Liquid |