missing translation for 'onlineSavingsMsg'
Learn More

golgin 97 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 12640-1-AP-150UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16840464

  • 381.34 CHF / 150 Mikroliter
Verfügbar ab: 17-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The MW of this protein is 97 kDa, and 12640-1-AP specifically recognizes the 97 kDa protein. IF result shows strong Golgi staining.

The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. The Golgins are a family of proteins, of which the protein encoded by this gene is a member, that are localized to the Golgi. This encoded protein is associated with Sjogren′s syndrome.
Spezifikation

Spezifikation

golgin 97
Polyclonal
Unconjugated
GOLGA1
GOLGA1, Golgin 97, Golgin subfamily A member 1
Rabbit
Antigen Affinity Chromatography
RUO
2800, 76899
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q92805, Q9CW79
Golga1
Golgin 97 Fusion Protein Ag3335
150 μL
Primary
Human, Mouse
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote