missing translation for 'onlineSavingsMsg'
Learn More
Learn More
golgin 97 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | golgin 97 |
---|---|
Konzentration | 0.13 mg/mL |
Anwendungen | Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16850464
|
Proteintech
12640-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 07-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16840464
|
Proteintech
12640-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 07-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The MW of this protein is 97 kDa, and 12640-1-AP specifically recognizes the 97 kDa protein. IF result shows strong Golgi staining.
The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. The Golgins are a family of proteins, of which the protein encoded by this gene is a member, that are localized to the Golgi. This encoded protein is associated with Sjogren′s syndrome.Spezifikation
golgin 97 | |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q92805, Q9CW79 | |
2800, 76899 | |
Golgin 97 Fusion Protein Ag3335 | |
Primary | |
-20°C | |
GOLGA1 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
GOLGA1, Golgin 97, Golgin subfamily A member 1 | |
Golga1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |