missing translation for 'onlineSavingsMsg'
Learn More

Gold-Nanopartikel, 40 nm, Amin-funktionalisiert, 3 kDa PEGyliert, OD50, 530 nm-Absorption, Thermo Scientific™

Catalog Number p-7057613
Click to view available options
Menge:
0.5 mL
1 mL
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
7440-57-5
Au
196.97
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
23985
CHEBI:30050
Gold
[Au]
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
7440-57-5
Au
196.97
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
23985
CHEBI:30050
Gold
[Au]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Gold nanoparticles
Farbe Rosa bis Rot
Namenshinweis amine-functionalized, 3kDa PEGylated, OD: 50, 530nm absorption
Menge 0.5 mL
Partikelgröße 40nm
Empfindlichkeit Light sensitive
Form Flüssig
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'