missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rabbit IgG (H+L) Secondary Antibody, PE-Cyanine5.5, Invitrogen™
Goat Polyclonal Secondary Antibody
Marke: Invitrogen L42018
Additional Details : Gewicht : 0.50000kg
Beschreibung
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Spezifikation
Rabbit IgG (H+L) | |
Polyclonal | |
PBS with 0.1% sodium azide | |
Gamma Immunoglobins Heavy and Light chains | |
500 μL | |
Secondary | |
4° C, store in dark | |
IgG |
Flow Cytometry | |
PE-Cyanine5.5 | |
Goat | |
purified | |
RUO | |
Rabbit | |
Liquid |