missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, PE-Alexa Fluor™ 647, Invitrogen™
Goat Polyclonal Secondary Antibody
Marke: Invitrogen A20990
Additional Details : Gewicht : 0.10000kg
Beschreibung
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Spezifikation
Mouse IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
PE-Alexa Fluor 647 | |
Goat | |
Purified | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry | |
1 mg/mL | |
PBS with 1% glycerol, 2mM EDTA and 5mM sodium azide; pH 7.5 | |
Gamma Immunoglobins Heavy and Light chains | |
100 μg | |
Secondary | |
4° C, store in dark | |
IgG |