Learn More
Invitrogen™ Goat anti-Mouse IgG (H+L) Secondary Antibody, AP
Goat Polyclonal Secondary Antibody
Marke: Invitrogen™ 31321
Beschreibung
Product # 31321 has been successfully used in Western blot, IHC and IP applications. Product # 31321 reacts with the heavy chains of mouse IgG and with the light chains common to most mouse immunoglobulins, but does not react against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Reconstitute with 0.2 mL of distilled water (1 mg/mL after restoration).
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Spezifikation
| Mouse IgG (H+L) | |
| Polyclonal | |
| Alkaline Phosphatase (AP) | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| Mouse | |
| Secondary Antibody | |
| IgG |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 0.6 mg/mL | |
| TBS with 50mM sucrose, 15mg/mL BSA and no preservative; pH 8.0 | |
| Purified Mouse IgG, whole molecule. | |
| 200 μL | |
| Secondary | |
| 4°C | |
| Lyophilized |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.