Learn More
Goat anti-Mouse IgG (H+L) Secondary Antibody, DyLight™ 650, Invitrogen™
Goat Polyclonal Secondary Antibody
Marke: Invitrogen 84545
Additional Details : Gewicht : 2.72160kg
Beschreibung
Product # 84545 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 84545 reacts with the heavy chains of mouse IgG and with the light chains common to most mouse immunoglobulins, but does not react against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product protected from light at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. DyLight 650 Amax= 654 nm; Emax= 673 nm. Mole Dye/Mole Protein Ratio is lot-dependent.
Mouse IgG (H+L) Polyclonal antibody specifically detects Mouse IgG (H+L) in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot.Spezifikation
Mouse IgG (H+L) | |
Polyclonal | |
DyLight 650 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 1% BSA and 0.02% sodium azide; pH 7.2 | |
Purified Mouse IgG, whole molecule | |
1 mL | |
Secondary | |
4° C | |
IgG |