Learn More
Invitrogen™ Goat anti-Human IgG (Lambda light chain) Recombinant Secondary Antibody, HRP
Goat Recombinant Monoclonal Secondary Antibody
Marke: Invitrogen™ A5686650UL
Beschreibung
This Goat-anti-Human IgG (Lambda Light Chain), Recombinant Secondary Antibody specifically detects human IgG Lambda Light chain with no cross-reactivity to kappa light chain, heavy chain, and other species immunoglobulins. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include better specificity, animal origin-free formulation, and lot-to-lot consistency. The sensitivity and specificity of each lot is confirmed using ELISA. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Spezifikation
| Human IgG (Lambda light chain) | |
| Recombinant Monoclonal | |
| 1.0 mg/mL | |
| PBS with proprietary stabilizer and proprietary preservative; pH 7.4 | |
| Purified Human IgG. | |
| 50 μL | |
| Secondary | |
| 4°C, do not freeze | |
| Liquid |
| ELISA, Western Blot | |
| 85H45L7 | |
| HRP | |
| Goat | |
| Protein G | |
| RUO | |
| Human | |
| Secondary Antibody | |
| IgG |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.