missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Human IgG (Lambda Light Chain) Recombinant Secondary Antibody, Alexa Fluor™ Plus 488

Goat Recombinant Monoclonal Secondary Antibody

112.00 CHF - 272.00 CHF

Spezifikation

Antigen Human IgG (Lambda Light Chain)
Klon 85H45L7
Konzentration 1.0 mg/mL
Inhalt und Lagerung 4°C, store in dark
Anwendungen Flow Cytometry, Western Blot, Immunocytochemistry
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
17894584
missing translation for 'viewDocuments'
Invitrogen™
A56867
250 μL
272.00 CHF
250 Mikroliter
Alternative Produkte anzeigen
missing translation for 'toSaveUpto' 
»
om dit product te kopen Registreer vandaag om een webaccount aan te maken
17824584
missing translation for 'viewDocuments'
Invitrogen™
A5686750UL
50 μL
112.00 CHF
50 Mikroliter
Alternative Produkte anzeigen
missing translation for 'toSaveUpto' 
»
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung

Beschreibung

This Goat-anti-Human IgG (Lambda Light Chain), Recombinant Secondary Antibody specifically detects human IgG Lambda Light chain with no cross-reactivity to kappa light chain, heavy chain, and other species immunoglobulins. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include better specificity and lot-to-lot consistency. The sensitivity and specificity of each lot is confirmed using ELISA. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Spezifikation

Spezifikation

Human IgG (Lambda Light Chain)
1.0 mg/mL
Flow Cytometry, Western Blot, Immunocytochemistry
Alexa Fluor Plus 488
Goat
RUO
proprietary buffer with 0.008% Bromonitrodioxane, 0.008% Methylisothiazolone; pH 6.5
Secondary
Affinity chromatography
85H45L7
4°C, store in dark
Recombinant Monoclonal
Liquid
IgG
Human
Purified Human IgG.
Secondary Antibody
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt