Learn More
Goat anti-Human IgG Fc Secondary Antibody, DyLight 633, Novus Biologicals™
Goat Polyclonal Antibody
Marke: Novus Biologicals NBP1-75818
187.00 CHF gültig bis 2025-12-16
BESTER Aktionspreis! Benutzen Sie den Promotions-Code "24090" um Ihren Promotions-Preis zu erhalten.
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
IgG Fc Polyclonal antibody specifically detects IgG Fc in Human samples. It is validated for Flow Cytometry, Immunomicroscopy.
Spezifikation
| IgG Fc | |
| Polyclonal | |
| DyLight 633 | |
| O75015, O75015 | |
| Purified Human IgG, Fc fragment | |
| 1 mg | |
| Secondary | |
| Rehydrate with 1.1 ml of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily. | |
| Store lyophilized antibody at 4°C. Mix reconstituted liquid with an equal volume of glycerol, aliquot contents and freeze at -20°C or below for long term storage. |
| Flow Cytometry, Immunomicroscopy | |
| LYOPH | |
| Flow Cytometry 1:20 - 1:2000, Immunomicroscopy 1:20 - 1:2000 | |
| Goat | |
| Affinity purified | |
| RUO | |
| Based on IEP, this antibody reacts with heavy gamma chains on human IgG | |
| Human | |
| IgG |