missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human IgE Heavy Chain Secondary Antibody, DyLight 633, Novus Biologicals™
Goat Polyclonal Antibody
Marke: Novus Biologicals NBP1-75777
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
IgE Heavy Chain Polyclonal antibody specifically detects IgE Heavy Chain in Human samples. It is validated for Flow Cytometry, Immunomicroscopy.
Spezifikation
| IgE Heavy Chain | |
| Polyclonal | |
| DyLight 633 | |
| Constant Region of Heavy Chain of IgE, Ig Epsilon Chain C Region, Ig Epsilon Chain C Region ND, IgE, Immunoglobulin Epsilon, immunoglobulin heavy constant epsilon | |
| Goat | |
| Affinity purified | |
| RUO | |
| 3497 | |
| Rehydrate with 1.1 ml of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily. | |
| Store lyophilized antibody at 4°C. Mix reconstituted liquid with an equal volume of glycerol, aliquot contents and freeze at -20°C or below for long term storage. |
| Flow Cytometry, Immunomicroscopy | |
| LYOPH | |
| Flow Cytometry 1:20 - 1:2000, Immunomicroscopy 1:20 - 1:2000 | |
| IGHE | |
| Purified human IgE, (e chain) | |
| 1 mg | |
| Secondary | |
| Based on IEP, this antibody reacts with heavy epsilon chains on human IgE | |
| Human | |
| IgE |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur