missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Dog IgG F(c) fragment, Texas Red, Polyclonal, Secondary Antibody, Abnova™
Marke: Abnova PAB10533.2mg
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
| Goat anti-Dog IgG F(c) fragment | |
| Polyclonal | |
| Goat anti-dog IgG recognizes dog IgG F(c) fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Texas Red. | |
| Lyophilized from 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| Dog IgG F(c) fragment | |
| 2 mg | |
| Dog | |
| Lyophilized |
| Flow Cytometry, Immunofluorescence | |
| Texas Red | |
| The optimal working dilution should be determined by the end user. | |
| Goat | |
| This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. | |
| Secondary | |
| Store at 4°C on dry atmosphere. After reconstitution with 1 mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur