Learn More
Tocris Bioscience™ Go 6983
Artikelnummer. p-4783992
Click to view available options
Menge:
1 mg
10 mg
Packungsgröße:
1 Milligramm
10 Milligramm
Artikelnummer. 18074512
Marke: Tocris Bioscience™ 2285/1
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Broad spectrum PKC inhibitor
Broad spectrum protein kinase C (PKC) inhibitor (IC50 values are 7, 7, 6, 10, 60 and 20000 nM for PKCα, PKCβ, PKCγ, PKCδ, PKCζ and PKCμ respectively). Displays cardioprotective properties; reduces polymorphonuclear leukocyte adherence and infiltration following myocardial ischemia/reperfusion injury. Optimizes naïve human pluripotent stem cell growth and viability following naïve cell derivation from primed ESCs and iPSCs using naïve human stem cell medium (NHSM).
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 133053-19-7
Summenformel C26H26N4O3
Spezifikation
Chemischer Name oder Material Go 6983
Menge 1 mg
Formelmasse 442.51
Reinheit >98%
Molekulargewicht 442.519