Learn More
GNAI2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527520
Additional Details : Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: Jurkat. Predicted reactivity: Mouse (98%), Rat (99%), Japanese Medaka (90%), Xenopus laevis (95%), Dog (99%), Pig (100%), Chicken (97%), Sheep (99%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (99%), Guinea pig (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is an alpha subunit of guanine nucleotide binding proteins (G proteins). The encoded protein contains the guanine nucleotide binding site and is involved in the hormonal regulation of adenylate cyclase. Several transcript variants encoding different isoforms have been detected for this gene, but the full-length nature of only two are known so far.Spezifikation
GNAI2 | |
Polyclonal | |
Unconjugated | |
Gnai2 | |
adenylate cyclase-inhibiting G alpha protein; BR; C76432; CD49B; G alpha inhibitory type 2 (a); G alpha inhibitory type 2 (b); G protein subunit alpha i2; Galphai2; Gia; GIP; GNAI2; Gnai-2; GNAI2B; GPIa; GTP-binding protein (G-alpha-i2); GTP-binding regulatory protein Gi alpha-2 chain; guanidine nucleotide binding protein, (G protein) , alpha inhibiting activity polypeptide 2; guanine nucleotide binding protein (G protein), alpha inhibiting 2; guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 2; guanine nucleotide binding protein, alpha inhibiting 2; guanine nucleotide binding protein, alpha inhibiting activity polypeptide 2; guanine nucleotide-binding protein G(i) subunit alpha-2; guanine nucleotide-binding protein G(i), alpha-2 subunit; H_LUCA15.1; H_LUCA16.1; inhibitory GTP-binding protein subunit alpha 2; VLA-2; VLAA2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2771 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P04899 | |
Gnai2 | |
Recombinant fragment corresponding to a region within amino acids 94 and 355 of Human G protein alpha Inhibitor 2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |