missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GNA13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA552691
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Immunogen sequence: HGQDFDQRAR EEFRPTIYSN VIKGMRVLVD AREKLHIPWG DNSNQQHGDK MMSFDTRAPM AAQGMVETRV FLQYLPAIRA LWADSGIQNA YDRRREFQLG ES Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
Guanine nucleotide-binding proteins (G proteins) are involved as modulators or transducers in various transmembrane signaling systems (PubMed:15240885, PubMed:16787920, PubMed:16705036, PubMed:27084452). Activates effector molecule RhoA by binding and activating RhoGEFs (ARHGEF1/p115RhoGEF, ARHGEF11/PDZ-RhoGEF and ARHGEF12/LARG) (PubMed:15240885, PubMed:12515866). GNA13-dependent Rho signaling subsequently regulates transcription factor AP-1 (activating protein-1). Promotes tumor cell invasion and metastasis by activating RhoA/ROCK signaling pathway (PubMed:16787920, PubMed:16705036, PubMed:27084452). Inhibits CDH1-mediated cell adhesion in process independent from Rho activation (PubMed:11976333). [UniProt].
Spezifikation
| GNA13 | |
| Polyclonal | |
| Unconjugated | |
| GNA13 | |
| AU024132; AU043124; G alpha 13; G alpha-13; G protein subunit alpha 13; G13; Galpha 13; Galpha13; Gna13; Gna-13; G-protein subunit alpha-13; guanine nucleotide binding protein (G protein), alpha 13; guanine nucleotide binding protein, alpha 13; guanine nucleotide-binding protein subunit alpha-13 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10672 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14344 | |
| GNA13 | |
| Recombinant protein corresponding to Human GNA13. Recombinant protein control fragment (Product #RP-90637). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur