missing translation for 'onlineSavingsMsg'
Learn More

AcroBiosystems GMP Human IL-12 Protein

Artikelnummer. 31451662
Ansicht ändern
Click to view available options
Menge:
1 mg
10 mg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
31451662 1 mg
31453488 50 μg
31453189 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 31451662 Lieferant AcroBiosystems Lieferanten-Nr. GMPL12H231mg

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

IL-12 is produced by macrophages and B lymphocytes and has been shown to have multiple effects on T cells and natural killer (NK) cells. These effects include inducing production of IFN-gamma and TNF by resting and activated T and NK cells, synergizing with other IFN-gamma inducers at both the transcriptional and post-transcriptional levels. This interaction induces IFN-gamma gene expression, enhancing the cytotoxic activity of resting NK and T cells, inducing and synergizing with IL-2 in the generation of lymphokine-activated killer (LAK) cells, acting as a co-mitogen to stimulate proliferation of resting T cells, and inducing proliferation of activated T and NK cells. Current evidence indicates that IL‐12, produced by macrophages in response to infectious agents, is a central mediator of the cell‐mediated immune response by its actions on the development, proliferation, and activities of TH1 cells. In its role as the initiator of cell-mediated immunity, it has been suggested that IL-12 has therapeutic potential as a stimulator of cell-mediated immune responses to microbial pathogens, metastatic cancers, and viral infections such as AIDS.

Spezifikation

Zugriffsnummer Uniprot,Uniprot
Charakterisierung This protein carries no tag. The protein has a calculated MW of 58.9 kDa. The protein migrates as 76 kDa±3 kDa when calibrated against under reducing (R) condition (SDS-PAGE) due to glycosylation.
Format Powder-GMP
Zusammensetzung Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4 with protectants. Contact us for customized product form or formulation.
Qualitätskontrollen <0.5 ng/μg of protein tested by ELISA.∣<0.02 ng/μg of protein tested by qPCR.∣Less than 10 EU/mg, tested by the LAL method in compliance with USP 85 and Ph. Eur. 2.6.14.∣Sterility testing was performed using the membrane filtration method in compliance with USP &CM and Ph. Eur. 2.6.1.∣Negative
Menge 1 mg
Quelle GMP Human IL-12 Protein (GMP-L12H23) is expressed from human 293 cells (HEK293). It contains AA Ile 23 - Ser 328 & Arg 23 - Ser 219 (Accession # & ). Predicted N-terminus: Ile 23
Biologische Aktivität GMP Human IL-12 Protein (Cat. No. GMP-L12H23) stimulates secretion of IFN-γ by NK-92. The specific activity of GMP Human IL-12 Protein is >1.00x10^7 IU/mg, which is calibrated against human IL-12 WHO International Standard (NIBSC code: 95/544) (QC tested).∣The activity of GMP Human IL-12 Protein (Cat. No. GMP-L12H23) was higher than other competing products.∣Human PBMCs were cultured with GMP Human IL-2 Protein (ACROBiosystems, Cat. No. GMP-L02H14), GMP Human IL-12 Protein (ACROBiosystems, Cat. No. GMP-L12H23) and GMP Human 4-1BB Ligand Protein (ACROBiosystems, Cat. No. GMP-41LH26), in CelThreaTM GMP T Cell Expansion Medium (ACROBiosystems, Cat. No. GMP-CM3103) for two weeks. The result shows that GMP Human IL-12 Protein (ACROBiosystems) can promote the expansion of these cells with a reasonable cell viability, and can be comparable with the Competitor M IL-12 protein.∣Human PBMCs were cultured with GMP Human IL-2 Protein (ACROBiosystems, Cat. No. GMP-L02H14), GMP Human IL-12 Protein (ACROBiosystems, Cat. No. GMP-L12H23) and GMP Human 4-1BB Ligand Protein (ACROBiosystems, Cat. No. GMP-41LH26), in CelThreaTM GMP T Cell Expansion Medium (ACROBiosystems, Cat. No. GMP-CM3103) for two weeks. The result shows that GMP Human IL-12 Protein (ACROBiosystems) can be comparable with the Competitor M IL-12 protein.∣The Cell based assay shows that GMP Human IL-12 Protein (Cat. No. GMP-L12H23) is stable at 37°C for 24 hours.∣The Cell based assay shows that GMP Human IL-12 Protein (Cat. No. GMP-L12H23) is stable at 4°C for 180 days.∣The Cell based assay shows that GMP Human IL-12 Protein (Cat. No. GMP-L12H23) is stable after freezing and thawing 3 times.∣The Cell based assay shows batch-to-batch consistency between Acro's GMP and PG IL-12.
Spezies Human
Sterilität Sterile
Inhalt und Lagerung -20°C to -70°C for 5 years in lyophilized state; -70°C for 12 months under sterile conditions after reconstitution. Upon receipt, store it immediately at -20°C or lower for long term storage.
Endotoxin-Gehalt Less than 10 EU/mg, tested by the LAL method in compliance with USP 85 and Ph. Eur. 2.6.14.
Versandbedingung This product is supplied and shipped with blue ice, please inquire the shipping cost.
Spezifische Aktivität The activity of GMP Human IL-12 Protein (Cat. No. GMP-L12H23) was higher than other competing products.
Reinheits- oder Qualitätsgrad 0.95
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.