missing translation for 'onlineSavingsMsg'
Learn More

AcroBiosystems GMP Human IL-10 Protein

Artikelnummer. 31451649
Ansicht ändern
Click to view available options
Menge:
1 mg
10 mg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
31451649 1 mg
31451966 50 μg
31451673 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 31451649 Lieferant AcroBiosystems Lieferanten-Nr. GMPL10H251mg

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

The protein encoded by this gene is a cytokine produced primarily by monocytes and to a lesser extent by lymphocytes. This cytokine has pleiotropic effects in immunoregulation and inflammation. It down-regulates the expression of Th1 cytokines, MHC class II Ags, and costimulatory molecules on macrophages. It also enhances B cell survival, proliferation, and antibody production. This cytokine can block NF-kappa B activity, and is involved in the regulation of the JAK-STAT signaling pathway. Knockout studies in mice suggested the function of this cytokine as an essential immunoregulator in the intestinal tract. Mutations in this gene are associated with an increased susceptibility to HIV-1 infection and rheumatoid arthritis. [provided by RefSeq, May 2020]

Spezifikation

Zugriffsnummer Uniprot
Charakterisierung IL-10 Structure This protein carries no tag. The protein has a calculated MW of 18.6 kDa. The protein migrates as 17 kDa±3 kDa when calibrated against under reducing (R) condition (SDS-PAGE).
Format Powder-GMP
Zusammensetzung Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4 with protectants. Contact us for customized product form or formulation.
Qualitätskontrollen <0.5 ng/μg of protein tested by ELISA.∣<0.02 ng/μg of protein tested by qPCR.∣Less than 10 EU/mg, tested by the LAL method in compliance with USP 85 and Ph. Eur. 2.6.14.∣Sterility testing was performed using the membrane filtration method in compliance with USP &CM and Ph. Eur. 2.6.1.∣Negative
Menge 1 mg
Quelle GMP Human IL-10 Protein (GMP-L10H25) is expressed from human 293 cells (HEK293). It contains AA Ser 19 - Asn 178 (Accession # ). Predicted N-terminus: Ser 19 Request for sequence
Biologische Aktivität GMP Human IL-10 Protein (Cat. No. GMP-L10H25) inhibits the secretion IL-6 by RAW264.7 cells. The specific activity of GMP Human IL-10 Protein is >1.00 x 10^7 IU/mg, which is calibrated against WHO Reference Reagent Interleukin-10 (Human, rDNA derived) (NIBSC code: 93/722) (QC tested).∣The activity of GMP Human IL-10 (Cat. No. GMP-L10H25) was higher than other competing products.∣The Cell based assay shows that GMP Human IL-10 (Cat. No. GMP-L10H25) is stable at 37 °C for 24 hours.∣The Cell based assay shows that GMP Human IL-10 Protein (Cat. No. GMP-L10H25) is stable at 4°C for 180 days.∣The Cell based assay shows that GMP Human IL-10 (Cat. No. GMP-L10H25) is stable after freezing and thawing 3 times.∣The Cell based assay shows batch-to-batch consistency between Acro's GMP and PG IL-10.
Spezies Human
Sterilität Sterile
Inhalt und Lagerung -20°C to -70°C for 5 years in lyophilized state; -70°C for 12 months under sterile conditions after reconstitution. Upon receipt, store it immediately at -20°C or lower for long term storage.
Endotoxin-Gehalt Less than 10 EU/mg, tested by the LAL method in compliance with USP 85 and Ph. Eur. 2.6.14.
Versandbedingung This product is supplied and shipped with blue ice, please inquire the shipping cost.
Spezifische Aktivität The activity of GMP Human IL-10 (Cat. No. GMP-L10H25) was higher than other competing products.
Reinheits- oder Qualitätsgrad 0.95
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.