Learn More
GLYCTK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12426-1-AP-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This locus encodes a member of the glycerate kinase type-2 family. The encoded enzyme catalyzes the phosphorylation of (R)-glycerate and may be involved in serine degradation and fructose metabolism. Decreased activity of the encoded enzyme may be associated with the disease D-glyceric aciduria. Alternatively spliced transcript variants have been described.Spezifikation
| GLYCTK | |
| Polyclonal | |
| Unconjugated | |
| GLYCTK | |
| glycerate kinase, GLYCTK, HBeAg binding protein 4, HBeAgBP4A, HBEBP2, HBEBP4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 132158, 235582, 684314 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q0VGK3, Q8IVS8, Q8QZY2 | |
| Glyctk | |
| GLYCTK Fusion Protein Ag3097 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |