missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLIPR1L1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20828-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The function of this protein remains unknown.Spezifikation
GLIPR1L1 | |
Polyclonal | |
Unconjugated | |
GLIPR1L1 | |
ALKN2972, GLIPR1 like protein 1, GLIPR1L1, PRO7434 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
256710 | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6UWM5 | |
GLIPR1L1 | |
GLIPR1L1 Fusion Protein Ag14852 | |
150 μL | |
Primary | |
Human | |
Liquid |