missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human CCL15 (MIP-5) Recombinant Protein

Human CCL15 (MIP-5) Recombinant Protein

Marke:  Gibco™ PHC1544

Additional Details : Gewicht : 0.50000kg

Artikelnummer. 10757853

  • 1056.00 CHF / 1 Stück
Verfügbar ab: 03-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Human CCL15 (MIP-5) Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN

MIP-5 (Macrophage inflammatory protein-5 also known as CCL15 and MIP-1δ) is a chemoattractant for monocytes, T-lymphocytes and eosinophils.

Shipping Condition: Wet Ice.

Spezifikation

Spezifikation

Cell Culture
10.1 kDa
25 ug
92 aa
Store in freezer (-5 to -30°C).
< 0.1 ng/μg
Wet Ice
Chemokines & Receptors
Lyophilized
Immunology, Inflammation, Signal Transduction, Neurobiology
98 %
6359
Sequential Chromatography
MIP-5
Human
Carrier-Free
25 μg
E. coli
Recombinant, Ligand
MIP (Macrophage Inflammatory Protein) Chemokines
Gibco™
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only. Not for use in diagnostic procedures.